4'-Methoxyacetophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

15236-0010 15236-2500
TS15236-0010EA 130 USD
TS15236-0010 TS15236-2500
4'-Methoxyacetophenone 98%
4'-Methoxyacetophenone
Formula: C₉H₁₀O₂
Boiling Pt: 152 °C (1013 hPa)
Melting Pt: 36…39 °C
Flash Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00008745
CAS Number: 100-06-1
EINECS: 202-815-9

Order Now

Specification Test Results

Appearance White Crystalline mass
Infrared spectrum Conforms
Melting point 36 to 40 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR