ω-Bromoacetophenone 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: α-Bromoacetophenone, Phenacyl bromide, 2-Bromoacetophenone

15201-0250 15201-1000 15201-5000
TS15201-0250EA 45 USD
TS15201-0250 TS15201-1000 TS15201-5000
ω-Bromoacetophenone 98%
ω-Bromoacetophenone
Formula: (C₆H₅)C(O)CH₂Br
MW: 199.05 g/mol
Boiling Pt: 135 °C (18 mmHg)
Melting Pt: 46…51 °C
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00000195
CAS Number: 70-11-1
EINECS: 200-724-9
UN: 2645
ADR: 6.1,II
Merck Index: 13,01386

Order Now

Specification Test Results

Appearance White to green-brown Powder or crystals
Infrared spectrum Conforms
Melting point 46 to 51 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR