ω-Bromo-3-methoxyacetophenone 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
15191-0100
15191-0250
TS15191-0100EA
96
USD
TS15191-0100
TS15191-0250
ω-Bromo-3-methoxyacetophenone 97%
ω-Bromo-3-methoxyacetophenone
Formula:
C₉H₉BrO₂ Boiling Pt: 285 °C (1013 hPa) Melting Pt: 59…64 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000199 CAS Number: 5000-65-7 EINECS: 225-666-1 UN: 3261 ADR: 8,II |
Specification Test Results
Appearance | White to yellow-green Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 59 to 64 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...