Hexanal 96% stabilised

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Hexylaldehyde, Caproaldehyde, Hexaldehyde, Aldehyde C6, Hexyl aldehyde

15185-0010 15185-0025 15185-5000
TS15185-0010EA 174 USD
TS15185-0010 TS15185-0025 TS15185-5000
Hexanal 96% stabilised
Hexanal
Formula: C₆H₁₂O
Boiling Pt: 130…131 °C (760 mmHg)
Melting Pt: –56 °C
Density: 0.815 g/cm³
Flash Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00007027
CAS Number: 66-25-1
EINECS: 200-624-5
UN: 1207
ADR: 3,III
Merck Index: 13,01766

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4020 to 1.4070 (20 °C, 589 nm)
Density (20°C) 0.810 to 0.820 g/ml
GC ≥95 %
Stabilizer 100 to 500 ppm alpha-Tocopherol
Acid value ≤10 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR