4-(4-Nitrophenylazo)resorcinol 90%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-((4-Nitrophenyl)diazenyl)benzene-1,3-diol, Azoviolet, Azo Violet, Magneson I
15151-0250
15151-1000
TS15151-0250EA
58
USD
TS15151-0250
TS15151-1000
4-(4-Nitrophenylazo)resorcinol 90%
4-(4-Nitrophenylazo)resorcinol
Formula:
C₁₂H₉N₃O₄ Melting Pt: 185 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00007310 CAS Number: 74-39-5 EINECS: 200-808-5 |
Specification Test Results
Appearance | Orange-red to red Powder |
Infrared spectrum | Conforms |
HPLC | ≥88 % |
Specific extinction E (1%/1cm) | ≥1400 (554 to 559 nm, 1N NaOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...