Isatin 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
2,3-Indolinedione, Indoline-2,3-dione, 1H-Indole-2,3-dione
15149-1000
15149-5000
TS15149-1000EA
46
USD
TS15149-1000
TS15149-5000
Isatin 98%
Isatin
Formula:
C₈H₅NO₂ MW: 147.13 g/mol Boiling Pt: 342 °C (1013 hPa) Melting Pt: 201…204 °C Flash Pt: 220 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005718 CAS Number: 91-56-5 EINECS: 202-077-8 Merck Index: 13,05119 |
Specification Test Results
Appearance | Orange to red Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 199 to 204 °C |
Water | ≤1.0 % (K.F.) |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...