Crotonic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: trans-Crotonic acid, 2-Butenoic acid, trans-2-Butenoic acid

15087-0025 15087-5000
TS15087-0025EA 143 USD
TS15087-0025 TS15087-5000
Crotonic acid 98%
Crotonic acid
Formula: C₄H₆O₂
MW: 86.09 g/mol
Boiling Pt: 185 °C (760 mmHg)
Melting Pt: 72 °C
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00002701
CAS Number: 107-93-7
EINECS: 203-533-9
UN: 2823
ADR: 8,III
Merck Index: 13,02625

Order Now

Specification Test Results

Appearance White to light yellow Crystalline flakes
Infrared spectrum Conforms
Water ≤0.7 %
GC ≥97.5 %
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR