Diethyl succinate 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Succinic acid diethyl ester, 1,4-Butanedioic acid diethyl ester
15054-0010
15054-0051
15054-0250
TS15054-0010EA
63
USD
TS15054-0010
TS15054-0051
TS15054-0250
Diethyl succinate 98%
Diethyl succinate
Formula:
C₈H₁₄O₄ MW: 174.2 g/mol Boiling Pt: 217 °C (760 mmHg) Melting Pt: –20 °C Density: 1.04 g/cm³ Flash Pt: 90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009208 CAS Number: 123-25-1 EINECS: 204-612-0 Merck Index: 12,09038 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4180 to 1.4220 (20 °C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...