2-Methyl-2-propen-1-ol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
15029-0100
15029-1000
15029-5000
TS15029-0100EA
76
USD
TS15029-0100
TS15029-1000
TS15029-5000
2-Methyl-2-propen-1-ol 98%
2-Methyl-2-propen-1-ol
Formula:
C₄H₈O Boiling Pt: 113…115 °C (760 mmHg) Melting Pt: –50 °C Density: 0.85 g/cm³ Flash Pt: 33 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004737 CAS Number: 513-42-8 EINECS: 208-161-0 UN: 2614 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4250 to 1.4270 (20 °C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...