4-Penten-1-ol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
15001-0100
15001-0500
TS15001-0100EA
120
USD
TS15001-0100
TS15001-0500
4-Penten-1-ol 99%
4-Penten-1-ol
Formula:
C₅H₁₀O Boiling Pt: 137 °C (1013 hPa) Density: 0.834 g/cm³ Flash Pt: 43 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002975 CAS Number: 821-09-0 EINECS: 212-473-2 UN: 1987 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4289 to 1.4309 (20 °C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...