2,6-Dimethylcyclohexanol (mixture of isomers) 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,6-Dimethylcyclohexanol
14981-0050
14981-0500
TS14981-0050EA
25
USD
TS14981-0050
TS14981-0500
2,6-Dimethylcyclohexanol (mixture of isomers) 99%
2,6-Dimethylcyclohexanol (mixture of isomers)
Formula:
C₈H₁₆O Density: 0.94 g/cm³ Flash Pt: 55 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001502 CAS Number: 5337-72-4 EINECS: 226-264-9 UN: 1987 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4585 to 1.4615 (20 °C, 589 nm) |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...