Dimethyl itaconate 97% stabilized
Supplier: Thermo Scientific Chemicals
Synonyms:
Itaconic acid dimethyl ester, Methylenesuccinic acid dimethyl ester
14916-0025
14916-1000
14916-5000
TS14916-0025EA
522
USD
TS14916-0025
TS14916-1000
TS14916-5000
Dimethyl itaconate 97% stabilized
Dimethyl itaconate
Formula:
C₇H₁₀O₄ MW: 158.15 g/mol Boiling Pt: 208 °C (760 mmHg) Melting Pt: 34…39 °C Density: 1.24 g/cm³ Flash Pt: 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008446 CAS Number: 617-52-7 EINECS: 210-519-6 |
Specification Test Results
Appearance | White to light yellow Crystalline mass or liquid |
Infrared spectrum | Conforms |
Melting point | 34 to 39 °C |
GC | ≥96.0 % |
Stabilizer | 100 ppm hydroquinone |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...