Cyclohexanecarboxaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

14867-0250 14867-1000
TS14867-0250EA 80 USD
TS14867-0250 TS14867-1000
Cyclohexanecarboxaldehyde 97%
Cyclohexanecarboxaldehyde
Formula: C₇H₁₂O
MW: 112.17 g/mol
Boiling Pt: 161…163 °C (760 mmHg)
Density: 0.926 g/cm³
Flash Pt: 41 °C
Storage Temperature: Ambient
MDL Number: MFCD00001457
CAS Number: 2043-61-0
UN: 1989
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light pink Liquid
Infrared spectrum Conforms
Refractive index 1.4490 to 1.4520 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR