Methyl-1-methylcyclopropyl ketone 95%

Supplier: Thermo Scientific Chemicals
Warning

14763-0050 14763-0250
TS14763-0050EA 75 USD
TS14763-0050 TS14763-0250
Methyl-1-methylcyclopropyl ketone 95%
Methyl-1-methylcyclopropyl ketone
Formula: C₆H₁₀O
Boiling Pt: 125…128 °C (760 mmHg)
Density: 0.895 g/cm³
Flash Pt: 24 °C
Storage Temperature: Ambient
MDL Number: MFCD00010245
CAS Number: 1567-75-5
UN: 1224
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4320 to 1.4350 (20 °C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR