2-(Bromoacetyl)naphthalene 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: α-Bromo-2'-acetonaphthone, 2-Bromo-2'-acetonaphthone

14748-0100 14748-0500
TS14748-0100EA 73 USD
TS14748-0100 TS14748-0500
2-(Bromoacetyl)naphthalene 98%
2-(Bromoacetyl)naphthalene
Formula: C₁₂H₉BrO
MW: 249.11 g/mol
Melting Pt: 82…85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004109
CAS Number: 613-54-7
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance Brown-purple Crystalline powder
Infrared spectrum Conforms
Melting point 82 to 85 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR