ω-Bromo-4-phenylacetophenone 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Bromo-4'-phenylacetophenone, Bromomethyl p-phenylphenyl ketone, α-Bromo-4-phenylacetophenone, Biphenyl-4-yl bromomethyl ketone, 4-Phenylphenacyl bromide

14635-0050 14635-0250
TS14635-0050EA 33 USD
TS14635-0050 TS14635-0250
ω-Bromo-4-phenylacetophenone 98%
ω-Bromo-4-phenylacetophenone
Formula: C₁₄H₁₁BrO
MW: 275.14 g/mol
Boiling Pt: 354 °C (1013 hPa)
Melting Pt: 123…127 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000202
CAS Number: 135-73-9
EINECS: 205-217-6
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance Yellow to beige to light brown Powder
Infrared spectrum Conforms
Melting point 123 to 127 °C
GC ≥97.5 %
Specific optical rotation -3.0° to -5.0° (20 °C, 589 nm) (c=2 in methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR