ω-Bromo-2-methoxyacetophenone 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Methoxyphenacyl bromide, α-Bromo-2-methoxyacetophenone, 2-Bromo-2'-methoxyacetophenone, 2-Bromo-1-(2-methoxyphenyl)ethanone, 2-Bromo-1-(2-methoxyphenyl)ethan-1-one, Bromomethyl o-methoxyphenyl ketone
14627-0010
14627-0050
14627-0250
TS14627-0010EA
43
USD
TS14627-0010
TS14627-0050
TS14627-0250
ω-Bromo-2-methoxyacetophenone 98%
ω-Bromo-2-methoxyacetophenone
Formula:
C₉H₉BrO₂ MW: 229.07 g/mol Boiling Pt: 130 °C (1 mmHg) Melting Pt: 43…45 °C Flash Pt: 113 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000196 CAS Number: 31949-21-0 UN: 3261 ADR: 8,II |
Specification Test Results
Appearance | White to brown Crystalline powder or solid |
Infrared spectrum | Conforms |
Melting point | 42 to 47 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...