Valerophenone 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-Phenyl-1-pentanone, 1-Phenylpentan-1-one

14080-0050 14080-0250 14080-1000 14080-5000
TS14080-0050EA 19 USD
TS14080-0050 TS14080-0250 TS14080-1000 TS14080-5000
Valerophenone 98%
Valerophenone
Formula: C₆H₅C(O)(CH₂)₃CH₃
MW: 162.23 g/mol
Boiling Pt: 105…107 °C (5 mmHg)
Melting Pt: –9 °C
Density: 0.98 g/cm³
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00009480
CAS Number: 1009-14-9
EINECS: 213-767-3

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5130 to 1.5170 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR