2-Undecanone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
14069-0010
14069-0050
14069-2500
TS14069-0010EA
222
USD
TS14069-0010
TS14069-0050
TS14069-2500
2-Undecanone 98%
2-Undecanone
Formula:
C₁₁H₂₂O Boiling Pt: 231…232 °C (760 mmHg) Melting Pt: 11…13 °C Density: 0.824 g/cm³ Flash Pt: 88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009583 CAS Number: 112-12-9 EINECS: 203-937-5 UN: 3082 ADR: 9,III Merck Index: 12,06178 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4280 to 1.4310 (20 °C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...