1-Undecanol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Undecyl alcohol, Undecyl alcohol, Undecan-1-ol, Decyl carbinol, 1-Hydroxy undecane
14068-1000
14068-5000
TS14068-1000EA
77
USD
TS14068-1000
TS14068-5000
1-Undecanol 98%
1-Undecanol
Formula:
C₁₁H₂₄O MW: 172.31 g/mol Boiling Pt: 146 °C (30 mmHg) Melting Pt: 11 °C Density: 0.831 g/cm³ Flash Pt: >112 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004751 CAS Number: 112-42-5 EINECS: 203-970-5 UN: 3082 ADR: 9,III |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4370 to 1.4420 (20 °C, 589 nm) |
GC | ≥97.5 % |
Specific gravity | (25/25°C) 0.829 to 0.833 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...