1,1,1-Tris(hydroxymethyl)ethane 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(Hydroxymethyl)-2-methyl-1,3-propanediol, Ethylidynetrimethanol

14051-0025 14051-0250 14051-5000
TS14051-0025EA 150 USD
TS14051-0025 TS14051-0250 TS14051-5000
1,1,1-Tris(hydroxymethyl)ethane 97%
1,1,1-Tris(hydroxymethyl)ethane
Formula: C₅H₁₂O₃
MW: 120.15 g/mol
Melting Pt: 200…203 °C
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00004687
CAS Number: 77-85-0
EINECS: 201-063-9

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR