1,1,1-Trifluoroacetone 98+%

Supplier: Thermo Scientific Chemicals
Danger

13974-0250 13974-1000 13974-5000
TS13974-0250EA 123 USD
TS13974-0250 TS13974-1000 TS13974-5000
1,1,1-Trifluoroacetone 98+%
1,1,1-Trifluoroacetone
Formula: C₃H₃F₃O
Boiling Pt: 22 °C (760 mmHg)
Melting Pt: –78 °C
Density: 1.25 g/cm³
Flash Pt: –30 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000423
CAS Number: 421-50-1
EINECS: 207-005-9
UN: 2924
ADR: 3,I

Order Now

Specification Test Results

Appearance Colorless to yellow to orange Liquid
Infrared spectrum Conforms
GC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR