Tiglic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (E)-2-Methylbut-2-enoic acid, trans-2,3-Dimethylacrylic acid

13897-0250 13897-1000 13897-5000
TS13897-0250EA 38 USD
TS13897-0250 TS13897-1000 TS13897-5000
Tiglic acid 98%
Tiglic acid
Formula: C₅H₈O₂
MW: 100.12 g/mol
Melting Pt: 61…65 °C
Density: 0.96 g/cm³
Flash Pt: 95 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00066864
CAS Number: 80-59-1
EINECS: 201-295-0
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White to beige Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 61 to 65 °C
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR