Thiophene-2-carbaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Thiophenecarboxaldehyde, Thiophene-2-carboxaldehyde

13883-0010 13883-0250 13883-1000
TS13883-0010EA 412 USD
TS13883-0010 TS13883-0250 TS13883-1000
Thiophene-2-carbaldehyde 98%
Thiophene-2-carbaldehyde
Formula: C₅H₄OS
MW: 112.15 g/mol
Boiling Pt: 198 °C (760 mmHg)
Density: 1.2 g/cm³
Flash Pt: 77 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005429
CAS Number: 98-03-3
EINECS: 202-629-8

Order Now

Specification Test Results

Appearance Clear yellow to light brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5890 to 1.5920 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR