1-(2-Thenoyl)-3,3,3-trifluoroacetone 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (TTA), Thenoyltrifluoroacetone, 4,4,4-Trifluoro-1-(2-thienyl)butane-1,3-dione, TTFA, 2-Thenoyltrifluoroacetone, (2-Thenoyl)-ω,ω,ω-trifluoroacetone

13861-0250 13861-1000 13861-5000
TS13861-0250EA 68 USD
TS13861-0250 TS13861-1000 TS13861-5000
1-(2-Thenoyl)-3,3,3-trifluoroacetone 99%
1-(2-Thenoyl)-3,3,3-trifluoroacetone
Formula: C₈H₅F₃O₂S
MW: 222.19 g/mol
Boiling Pt: 96…98 °C (8 mmHg)
Melting Pt: 40…44 °C
Flash Pt: 111 °C
Storage Temperature: Ambient
MDL Number: MFCD00005445
CAS Number: 326-91-0
EINECS: 206-316-7

Order Now

Specification Test Results

Appearance Light yellow to beige to brown Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 40 to 44 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR