3-Methoxy-1-propanol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Trimethylene glycol monomethyl ether

13715-0100 13715-0500
TS13715-0100EA 61 USD
TS13715-0100 TS13715-0500
3-Methoxy-1-propanol 98%
3-Methoxy-1-propanol
Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 149…150 °C (760 mmHg)
Density: 0.942 g/cm³
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00039550
CAS Number: 1589-49-7
UN: 1987
ADR: 3 III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4120 to 1.4140 (20 °C, 589 nm)
GC ≥97.5 %
Specific optical rotation +8° to +11° (20°C, 589 nm) (c=1,CH3OH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR