Syringaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

13288-0050 13288-0250 13288-1000
TS13288-0050EA 22 USD
TS13288-0050 TS13288-0250 TS13288-1000
Syringaldehyde 98%
Syringaldehyde
Formula: C₉H₁₀O₄
Boiling Pt: 192…193 °C (14 mmHg)
Melting Pt: 110…113 °C
Density: 1.013 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00006943
CAS Number: 134-96-3
EINECS: 205-167-5
Merck Index: 12,09191

Order Now

Specification Test Results

Appearance Light yellow to beige to green to brown Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 110 to 114 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR