2-Pyrrolecarbaldehyde 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1H-Pyrrole-2-carboxaldehyde, Pyrrole-2-carboxaldehyde, 1H-Pyrrole-2-carbaldehyde, Pyrrole-2-carbaldehyde

13206-0050 13206-0250 13206-1000
TS13206-0050EA 60 USD
TS13206-0050 TS13206-0250 TS13206-1000
2-Pyrrolecarbaldehyde 99%
2-Pyrrolecarbaldehyde
Formula: C₅H₅NO
MW: 95.1 g/mol
Melting Pt: 43…47 °C
Flash Pt: 106 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005217
CAS Number: 1003-29-8
EINECS: 213-705-5

Order Now

Specification Test Results

Appearance White to brown Low melting crystalline solid (May darken on storage)
Infrared spectrum Conforms
Melting point 40 to 46 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR