3-Pyridylmethanol 98%

Supplier: Thermo Scientific Chemicals
Warning

13200-0050 13200-1000
TS13200-0050EA 31 USD
TS13200-0050 TS13200-1000
3-Pyridylmethanol 98%
3-Pyridylmethanol
Formula: C₆H₇NO
Boiling Pt: 259 °C (760 mmHg)
Melting Pt: –7 °C
Density: 1.124 g/cm³
Flash Pt: 149 °C
Storage Temperature: Ambient
MDL Number: MFCD00006407
CAS Number: 100-55-0
EINECS: 202-864-6
Merck Index: 12,06614

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5425 to 1.5455 (20 °C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR