Isonicotinaldehyde 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Pyridine-4-carboxaldehyde, 4-Pyridinecarboxaldehyde, 4-Pyridinecarbaldehyde

13184-0250 13184-1000 13184-5000
TS13184-0250EA 54 USD
TS13184-0250 TS13184-1000 TS13184-5000
Isonicotinaldehyde 98%
Isonicotinaldehyde
Formula: C₆H₅NO
MW: 107.11 g/mol
Boiling Pt: 77…78 °C (12 mmHg)
Density: 1.138 g/cm³
Flash Pt: 82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006425
CAS Number: 872-85-5
EINECS: 212-832-3
UN: 1760
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5420 to 1.5450 (20 °C, 589 nm)
GC ≥97.5 %
Chloride (Cl) ≤0.003 %
Iron (Fe) ≤0.002 %
Heavy metals (as Pb) ≤0.002 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR