(±)-1-Phenyl-1,2-ethanediol 97%

Supplier: Thermo Scientific Chemicals

Synonyms: (±)-α,β-Dihydroxyethylbenzene, (±)-Phenylethane-1,2-diol, 1-Phenylethane-1,2-diol

13059-0050 13059-0250 13059-1000
TS13059-0050EA 34 USD
TS13059-0050 TS13059-0250 TS13059-1000
(±)-1-Phenyl-1,2-ethanediol 97%
(±)-1-Phenyl-1,2-ethanediol
Formula: C₆H₅CH(OH)CH₂OH
MW: 138.17 g/mol
Boiling Pt: 272…274 °C (760 mmHg)
Melting Pt: 61…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00003546
CAS Number: 93-56-1
EINECS: 202-258-1
Merck Index: 12,09029

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 61 to 67 °C
GC ≥99.0 %
UV at 310 nm A: ≤0.01
Relative density (20/20 °C) 0.785 to 0.789
Specific gravity (25 °C) 0.783 to 0.787
Identification D Passes test
Non-volatile matter ≤20 ppm
Related substances ≤0.3 %
Acidity/alkalinity Passes test
Benzene ≤2 ppm
Peroxides Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR