2-Phenylethanol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Phenethyl alcohol, Benzylcarbinol, β-Hydroxyethylbenzene, Benzyl carbinol, Benzenethanol

13018-0010 13018-0025 13018-0050 13018-0100 13018-1000 13018-2500
TS13018-0010EA 129 USD
TS13018-0010 TS13018-0025 TS13018-0050 TS13018-0100 TS13018-1000 TS13018-2500
2-Phenylethanol 99%
2-Phenylethanol
Formula: C₆H₅CH₂CH₂OH
MW: 122.17 g/mol
Boiling Pt: 219 °C (760 mmHg)
Melting Pt: –27 °C
Density: 1.02 g/cm³
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00002886
CAS Number: 60-12-8
EINECS: 200-456-2
Merck Index: 12,07370

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5300 to 1.5350 (20 °C, 589 nm)
GC ≥98.5 %
Specific gravity (25/25°C) 1.016 to 1.021
UV at 260 nm A: ≤0.02
Titration after Ion exchange ≥99.0 % (on dry substance)
Membrane filtration test (on 0.2 M soln.) Passes test
Suitability (HPLC gradient) Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR