(±)-1-Octen-3-ol 98%

Supplier: Thermo Scientific Chemicals
Warning

12947-0050 12947-0250 12947-1000
TS12947-0050EA 25 USD
TS12947-0050 TS12947-0250 TS12947-1000
(±)-1-Octen-3-ol 98%
(±)-1-Octen-3-ol
Formula: C₈H₁₆O
Boiling Pt: 174 °C (760 mmHg)
Density: 0.837 g/cm³
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00004589
CAS Number: 3391-86-4
EINECS: 222-226-0

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4340 to 1.4420 (20 °C, 589 nm)
Density (20°C) 0.832 to 0.842 kg/l
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR