(±)-2-Octanol 97%

Supplier: Thermo Scientific Chemicals
Danger

12941-0010 12941-0025 12941-1000
TS12941-0010EA 58 USD
TS12941-0010 TS12941-0025 TS12941-1000
(±)-2-Octanol 97%
(±)-2-Octanol
Formula: C₈H₁₈O
Boiling Pt: 174…181 °C (760 mmHg)
Melting Pt: –38 °C
Density: 0.81 g/cm³
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00004591
CAS Number: 123-96-6
EINECS: 204-667-0
Merck Index: 13,06785

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4250 to 1.4270 (20 °C, 589 nm)
GC ≥96.0 %
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR