1-Octadecanol 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Stearyl alcohol, 1-Hydroxyoctadecane, Heptadecyl carbinol, 1-Octadecyl alcohol

12930-0010 12930-0025
TS12930-0010EA 61 USD
TS12930-0010 TS12930-0025
1-Octadecanol 95%
1-Octadecanol
Formula: H₃C(CH₂)₁₆CH₂OH
MW: 270.5 g/mol
Boiling Pt: 336
Melting Pt: 55…58 °C
Flash Pt: 185 °C
Storage Temperature: Ambient
MDL Number: MFCD00002823
CAS Number: 112-92-5
EINECS: 204-017-6
Merck Index: 12,08960

Order Now

Specification Test Results

Appearance White Powder or flakes or pellets
Infrared spectrum Conforms
Melting point 55 to 60 °C
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR