Norcamphor 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Oxonorbornane, Bicyclo[2.2.1]heptan-2-one, Bicyclo[2.2.1]-2-heptanone, 2,5-Methanocyclohexanone, 2-Norbornanone

12925-0050 12925-0250 12925-1000
TS12925-0050EA 129 USD
TS12925-0050 TS12925-0250 TS12925-1000
Norcamphor 99%
Norcamphor
Formula: C₇H₁₀O
MW: 110.16 g/mol
Boiling Pt: 168…172 °C (760 mmHg)
Melting Pt: 88…91 °C
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00074823
CAS Number: 497-38-1
EINECS: 207-846-1
UN: 1325
ADR: 4.1,II

Order Now

Specification Test Results

Appearance Colorless to white Adhering crystals
Infrared spectrum Conforms
GC ≥98.5 %
UV at 320 nm A: ≤0.03
Color scale ≤10 APHA
Residue after evaporation ≤5 ppm
Titration with NaOH ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR