4-Nitrobenzonitrile 97%

Supplier: Thermo Scientific Chemicals
Danger

12848-0250 12848-1000
TS12848-0250EA 60 USD
TS12848-0250 TS12848-1000
4-Nitrobenzonitrile 97%
4-Nitrobenzonitrile
Formula: C₇H₄N₂O₂
Melting Pt: 144…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00007279
CAS Number: 619-72-7
EINECS: 210-610-0
UN: 3439
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 144 to 149 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR