4'-Nitroacetophenone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: p-Nitroacetophenone, 1-(4-Nitrophenyl)ethanone

12834-0010 12834-0050 12834-1000
TS12834-0010EA 278 USD
TS12834-0010 TS12834-0050 TS12834-1000
4'-Nitroacetophenone 97%
4'-Nitroacetophenone
Formula: C₈H₇NO₃
MW: 165.15 g/mol
Boiling Pt: 275
Melting Pt: 76…80 °C
Flash Pt: 149
Storage Temperature: Ambient
MDL Number: MFCD00007355
CAS Number: 100-19-6
EINECS: 202-827-4

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 76 to 80 °C
Water ≤0.6 %
GC ≥96.0 %
Free acid ≤0.2 % (as 4-Nitrobenzoic acid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR