3'-Nitroacetophenone 98%

Supplier: Thermo Scientific Chemicals

Synonyms: m-Nitroacetophenone

12833-1000 12833-5000
TS12833-1000EA 32 USD
TS12833-1000 TS12833-5000
3'-Nitroacetophenone 98%
3'-Nitroacetophenone
Formula: C₈H₇NO₃
MW: 165.15 g/mol
Boiling Pt: 202 °C (1013 hPa)
Melting Pt: 76…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00007259
CAS Number: 121-89-1
EINECS: 204-504-3

Order Now

Specification Test Results

Appearance Yellow to yellow-green to yellow-brown Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 76 to 80 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR