2'-Nitroacetophenone 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
o-Nitroacetophenone, 1-(2-Nitrophenyl)ethanone
12832-0025
12832-0200
TS12832-0025EA
34
USD
TS12832-0025
TS12832-0200
2'-Nitroacetophenone 95%
2'-Nitroacetophenone
Formula:
C₈H₇NO₃ MW: 165.15 g/mol Boiling Pt: 159 °C (16 mmHg) Melting Pt: 28…30 °C Flash Pt: >112 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00007144 CAS Number: 577-59-3 EINECS: 209-414-8 |
Specification Test Results
Appearance | Orange to brown Low melting solid |
Infrared spectrum | Conforms |
GC | ≥88 % |
Stabilizer | 0.05 % BHT |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...