1-Naphthaldehyde 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: α-Naphthaldehyde

12809-0025 12809-1000 12809-5000
TS12809-0025EA 1052 USD
TS12809-0025 TS12809-1000 TS12809-5000
1-Naphthaldehyde 95%
1-Naphthaldehyde
Formula: C₁₁H₈O
MW: 156.18 g/mol
Boiling Pt: 160…161 °C (15 mmHg)
Melting Pt: 1…2 °C
Density: 1.15 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004003
CAS Number: 66-77-3
EINECS: 200-633-4
UN: 3334
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear yellow to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.6500 to 1.6540 (20 °C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR