Mucobromic acid 99+%

Supplier: Thermo Scientific Chemicals
Danger

12805-1000 12805-5000
TS12805-1000EA 95 USD
TS12805-1000 TS12805-5000
Mucobromic acid 99+%
Mucobromic acid
Formula: C₄H₂Br₂O₃
Melting Pt: 122…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00063745
CAS Number: 488-11-9
EINECS: 207-670-5
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 122 to 125 °C
Titration with NaOH ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR