Monomethyl succinate 95%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Methoxy-4-oxobutanoic acid, Methyl hydrogen succinate, Succinic acid monomethyl ester

12773-0100 12773-0250 12773-1000
TS12773-0100EA 57 USD
TS12773-0100 TS12773-0250 TS12773-1000
Monomethyl succinate 95%
Monomethyl succinate
Formula: C₅H₈O₄
MW: 132.11 g/mol
Boiling Pt: 151 °C (20 mmHg)
Melting Pt: 55…59 °C
Storage Temperature: Ambient
MDL Number: MFCD00002788
CAS Number: 3878-55-5
EINECS: 223-408-2

Order Now

Specification Test Results

Appearance White to off-white Crystals and/or chunks
Infrared spectrum Conforms
Melting point 52 to 58 °C
GC ≥94 %
Sodium oxide (Na2O) ≤0.4 %
Silicon dioxide (SiO2) ≤0.03 %
Iron(III)oxide (Fe2O3) ≤0.03 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR