2-Formyl-6-picoline 97%

Supplier: Thermo Scientific Chemicals
Warning

12757-0010 12757-0050 12757-0250
TS12757-0010EA 66 USD
TS12757-0010 TS12757-0050 TS12757-0250
2-Formyl-6-picoline 97%
2-Formyl-6-picoline
Formula: C₇H₇NO
Melting Pt: 31…33 °C
Flash Pt: 68 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006291
CAS Number: 1122-72-1
EINECS: 214-359-8

Order Now

Specification Test Results

Appearance Light yellow to brown Low melting solid
Infrared spectrum Conforms
GC ≥96.0 %
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR