Methyl-2-hydroxyisobutyrate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 2-Hydroxyisobutyrate, 2-Hydroxyisobutyric Acid Methyl Ester

12697-0050 12697-0500
TS12697-0050EA 40 USD
TS12697-0050 TS12697-0500
Methyl-2-hydroxyisobutyrate 99%
Methyl-2-hydroxyisobutyrate
Formula: C₅H₁₀O₃
MW: 118.13 g/mol
Boiling Pt: 137 °C
Density: 1.03 g/cm³
Flash Pt: 41 °C
Storage Temperature: Ambient
MDL Number: MFCD00004457
CAS Number: 2110-78-3
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4100 to 1.4120 (20 °C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR