Methyl-5-chloro-5-oxovalerate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methyl glutaryl chloride, Methyl 4-chloroformylbutyrate

12654-0100 12654-0500
TS12654-0100EA 80 USD
TS12654-0100 TS12654-0500
Methyl-5-chloro-5-oxovalerate 98%
Methyl-5-chloro-5-oxovalerate
Formula: C₆H₉ClO₃
MW: 164.59 g/mol
Boiling Pt: 110 °C (17 mmHg)
Density: 1.191 g/cm³
Flash Pt: 82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000756
CAS Number: 1501-26-4
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4440 to 1.4470 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR