Methyl acetoacetate 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 3-oxobutanoate
12615-0010
12615-0025
12615-0100
12615-1000
TS12615-0010EA
56
USD
TS12615-0010
TS12615-0025
TS12615-0100
TS12615-1000
Methyl acetoacetate 99+%
Methyl acetoacetate
Formula:
C₅H₈O₃ Boiling Pt: 169…170 °C (760 mmHg) Melting Pt: –28 °C Density: 1.077 g/cm³ Flash Pt: 67 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008784 CAS Number: 105-45-3 EINECS: 203-299-8 Merck Index: 12,06090 |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4175 to 1.4200 (20 °C, 589 nm) |
GC | ≥99.0 % |
Color scale | ≤20 APHA |
Free acid (CH3COOH) | ≤0.2 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...