4'-Methoxypropiophenone 99%

Supplier: Thermo Scientific Chemicals
Warning

12609-0050 12609-0250 12609-1000
TS12609-0050EA 23 USD
TS12609-0050 TS12609-0250 TS12609-1000
4'-Methoxypropiophenone 99%
4'-Methoxypropiophenone
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 273…275 °C (760 mmHg)
Melting Pt: 27…29 °C
Density: 1.07 g/cm³
Flash Pt: 61 °C
Storage Temperature: Ambient
MDL Number: MFCD00009310
CAS Number: 121-97-1
EINECS: 204-512-7

Order Now

Specification Test Results

Appearance Clear colorless to amber Liquid
Infrared spectrum Conforms
Refractive index 1.5455 to 1.5475 (20 °C, 589 nm)
GC ≥98.5 %
Residue after evaporation ≤0.005 %
Acidity ≤0.0005 meq/g
Aluminium (Al) ≤0.5 ppm
Barium (Ba) ≤0.1 ppm
Boron (B) ≤0.02 ppm
Cadmium (Cd) ≤0.05 ppm
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.02 ppm
Cobalt (Co) ≤0.02 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Manganese (Mn) ≤0.02 ppm
Nickel (Ni) ≤0.02 ppm
Tin (Sn) ≤0.1 ppm
Zinc (Zn) ≤0.1 ppm
Peroxides ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR