5-Methoxy-2-methylindole 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Methoxy-2-methyl-1H-indole

12594-0010 12594-0050 12594-0250
TS12594-0010EA 36 USD
TS12594-0010 TS12594-0050 TS12594-0250
5-Methoxy-2-methylindole 99+%
5-Methoxy-2-methylindole
Formula: C₁₀H₁₁NO
MW: 161.2 g/mol
Melting Pt: 85…88 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005620
CAS Number: 1076-74-0
EINECS: 214-066-5

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 85 to 88 °C
GC ≥99.0 %
Elemental Analysis 14.80 to 18.83 % (S)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR