5-Methoxyindole 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Methoxy-1H-indole, Indol-5-yl methyl ether

12587-0050 12587-0250
TS12587-0050EA 97 USD
TS12587-0050 TS12587-0250
5-Methoxyindole 99%
5-Methoxyindole
Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 176…178 °C (17 mmHg)
Melting Pt: 52…56 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005674
CAS Number: 1006-94-6
EINECS: 213-745-3

Order Now

Specification Test Results

Appearance White to brown Powder
Infrared spectrum Conforms
Melting point 52 to 56 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR